missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Floating Centrifuge Tube Rack

Catalog No. 14127184
Change view
Click to view available options
Shape:
Round
Square
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Shape
14127184 Round
14127131 Square
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14127184 Supplier Fisherbrand™ Supplier No. HS2134AF2

For use on bench and in ice and water baths, floating microtube rack perfect for controlling sample temperature

  • The Floating Microtube Rack can be used on the bench, with water baths or ice containers. The rack keeps microtubes afloat in a water bath or can be pushed into crushed ice. It is perfect for controlling sample temperature
  • Made of polypropylene
  • Attach legs and rack stands upright for use on bench top
  • Detach legs and samples can be quickly removed by pushing down onto any surface

Specifications

Autoclavable Autoclavable
Color White
For Use With (Equipment) 1.5 or 2.0 Microtubes
Type Rack
Holds 20 Microtubes
Material Polypropylene
No. of Positions 20
No. per Pack 4
Shape Round
Diameter (Metric) Well 10.9 mm