Learn More
Labconco™ FlaskScrubber™ and SteamScrubber™ Glassware Washer Accessories
Make Labconco washers compatible with available electrical and plumbing
$1,063.11 - $1,734.11
| Catalog Number | Mfr. No. | Frequency | Voltage | Weight (English) Shipping | Weight (Metric) Shipping | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Frequency | Voltage | Weight (English) Shipping | Weight (Metric) Shipping | Price | Quantity & Availability | ||
|
10359209
|
Labconco™
4591901 |
50/60 Hz | 230 V | 8 lb. | 3.6 kg |
Each for $1,734.11
|
|
||
|
10359208
|
Labconco™
4591900 |
60 Hz | 115 V | 8 lb. | 3.6 kg |
Each for $1,734.11
|
|
||
|
10359206
|
Labconco™
4596100 |
60 Hz | 115 V | 60 lb. | 27.2 kg |
Each for $1,063.11
|
|
||
Description
Conversion Kits, Freestanding to Mobile
- Add mobility to freestanding SteamScrubber and FlaskScrubber Glassware Washers
- Not for use with undercounter washers or FlaskScrubber Vantage Series Glassware Washers
- Allow washers to be plugged into a 20A electrical wall outlet and connected to a standard laboratory sink faucet
- Kit consists of four 2 in. dia. rubber casters, two aerator adapters for connection to standard 55/64 in.-27 male and 15/16 in.-27 female laboratory faucets, drain hose and installation instructions
- For mixing cold tap water with hot drain water so that water <60°C (<140°F) is directed to house drain system
- Consist of plumbing components, solenoid valve and electrical connections
- When washer drainage pump is activated, solenoid valve opens, releasing flow of cold tap water to drain
Specifications
| Each |