Learn More
Labconco™ FlaskScrubber™ and SteamScrubber™ Glassware Washer Accessories
Make Labconco washers compatible with available electrical and plumbing
Supplier: Labconco™ 4596100
Description
Conversion Kits, Freestanding to Mobile
- Add mobility to freestanding SteamScrubber and FlaskScrubber Glassware Washers
- Not for use with undercounter washers or FlaskScrubber Vantage Series Glassware Washers
- Allow washers to be plugged into a 20A electrical wall outlet and connected to a standard laboratory sink faucet
- Kit consists of four 2 in. dia. rubber casters, two aerator adapters for connection to standard 55/64 in.-27 male and 15/16 in.-27 female laboratory faucets, drain hose and installation instructions
- For mixing cold tap water with hot drain water so that water <60°C (<140°F) is directed to house drain system
- Consist of plumbing components, solenoid valve and electrical connections
- When washer drainage pump is activated, solenoid valve opens, releasing flow of cold tap water to drain
Chemical Identifiers
| Each |
Specifications
| Freestanding to Mobile Conversion Kit | |
| Freestanding FlaskScrubber™, SteamScrubber™ glassware washer | |
| 60 lb. | |
| 115V 60Hz | |
| 60 Hz |
| Add mobility to freestanding SteamScrubber™ and FlaskScrubber™ glassware washers | |
| 10 ft. power cord with NEMA 5/20 plug, four 2 in. dia. rubber casters, two aerator adapters for laboratory faucets, drain hose and installation instructions | |
| 27.2 kg | |
| 115 V | |
| Each |