missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Flask Length PFTE Stopper

Stopper features a PTFE handle for easy gripping.
$48.82 - $176.18
Specifications
| Color | White |
|---|---|
| Material | PTFE |
| For Use With (Application) | Organic Synthesis |
| Quantity | 1 Ea. |
| Catalog Number | Mfr. No. | Stopper Number | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Stopper Number | Price | Quantity & Availability | |||||
31501900
![]() |
Synthware™
S120013 |
13 |
Each for $48.82
|
|
|||||
31501901
![]() |
Synthware™
S120016 |
16 |
Each for $55.61
|
|
|||||
31501902
![]() |
Synthware™
S120022 |
22 |
Each for $63.89
|
|
|||||
31501903
![]() |
Synthware™
S120027 |
27 |
Each for $94.57
|
|
|||||
31501904
![]() |
Synthware™
S120038 |
38 |
Each for $176.18
|
|
|||||
Specifications
| White | |
| PTFE | |
| Organic Synthesis | |
| 1 Ea. |