missing translation for 'onlineSavingsMsg'
Learn More

Synthware™ Flask Length PFTE Stopper
GREENER_CHOICE

Catalog No. 31501900
Change view
Click to view available options
Stopper Number:
13
16
22
27
38
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stopper Number
31501900 13
31501901 16
31501902 22
31501903 27
31501904 38
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 31501900 Supplier Synthware™ Supplier No. S120013

Stopper features a PTFE handle for easy gripping.

Specifications

Stopper Number 13
For Use With (Application) Organic Synthesis
Color White
Material PTFE
Quantity 1 Ea.
Size #13