Learn More
BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: Diol
FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance.
$543.04 - $2773.82
Specifications
Material Polypropylene
Type Diol
Resolution 40 to 60 μm
Column Type Diol
For Use With (Application) Alternate Selectivity for a Variety of Polar Types, Ideal for Lipids
View More Specs
Products 8
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity  
Catalog Number Mfr. No. Size (Metric) Tube Rack Includes Price Quantity  
05410046
View Documents
BUCHI
140000072
120 g 2 psc
Pack of 2 Each for $1,948.46
Only null left
05410042
View Documents
BUCHI
140000068
12 g 5 Pc.
Pack of 5 Each for $885.52
Only null left
05410043
View Documents
BUCHI
140000069
20 g 2 Pc.
Pack of 2 Each for $581.61
Only null left
05410047
View Documents
BUCHI
140000073
220 g 1 Pc.
Pack for $1,911.45
Only null left
05410048
View Documents
BUCHI
140000074
330 g 1 Pc.
Pack for $2,773.82
Only null left
05410044
missing translation for 'viewDocuments'
BUCHI
140000070
40 g 2 Pc.
Pack of 2 Each for $725.09
Only null left
05410041
missing translation for 'viewDocuments'
BUCHI
140000067
4 g 5 Pc.
Pack of 5 Each for $543.04
Only null left
05410045
missing translation for 'viewDocuments'
BUCHI
140000071
80 g 2 Pc.
Pack of 2 Each for $1,337.55
Only null left
Description
Description
  • Most economical solution without sacrificing quality for standard separations
  • Large selection of cartridge sizes from 4g-5,000g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
  • Silica is ideal for basic applications and polar compounds using organic solvents
  • Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
  • C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
  • Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
  • Neutral Alumina is ideal for aldehydes, ketones, and esters
Specifications
Specifications
Polypropylene
40 to 60 μm
Alternate Selectivity for a Variety of Polar Types, Ideal for Lipids
Diol
Diol
Any Flash System