Learn More
BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: Diol
FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance.
Supplier: BUCHI 140000071
Description
- Most economical solution without sacrificing quality for standard separations
- Large selection of cartridge sizes from 4g-5,000g
- Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
- Silica is ideal for basic applications and polar compounds using organic solvents
- Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
- C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
- Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
- Neutral Alumina is ideal for aldehydes, ketones, and esters
Specifications
| Polypropylene | |
| 2 Pc. | |
| 80 g | |
| Alternate Selectivity for a Variety of Polar Types, Ideal for Lipids |
| Diol | |
| 40 to 60 μm | |
| Diol | |
| Any Flash System |