missing translation for 'onlineSavingsMsg'
Learn More

BUCHI FlashPure EcoFlex Flash Chromatography Cartridges: Diol

Catalog No. 05410048
Change view
Click to view available options
Size (Metric) Tube Rack:
4 g
12 g
20 g
40 g
80 g
120 g
220 g
330 g
Includes:
1 Pc.
2 Pc.
2 psc
5 Pc.
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric) Tube Rack Includes
05410048 330 g 1 Pc.
05410046 120 g 2 psc
05410042 12 g 5 Pc.
05410043 20 g 2 Pc.
05410047 220 g 1 Pc.
05410044 40 g 2 Pc.
05410041 4 g 5 Pc.
05410045 80 g 2 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 05410048 Supplier BUCHI Supplier No. 140000074

FlashPure EcoFlex cartridges are designed to optimized backpressure, price and performance.

  • Most economical solution without sacrificing quality for standard separations
  • Large selection of cartridge sizes from 4g-5,000g
  • Non-RFID suitable on any system from BUCHI, ISCO, Biotage, Interchim or Yamazen
  • Silica is ideal for basic applications and polar compounds using organic solvents
  • Amino functionalized silica provides better peak shapes with polar and basic compounds and is ideal for carbohydrates
  • C18 functionalized silica is ideal for peptides and non-polar compounds in an aqueous mobile phase
  • Diol functionalized silica provides an alternate selectivity for a variety of polarity types and is ideal for lipids
  • Neutral Alumina is ideal for aldehydes, ketones, and esters

Specifications

Material Polypropylene
Type Diol
Includes 1 Pc.
Resolution 40 to 60 μm
Size (Metric) Tube Rack 330 g
Column Type Diol
For Use With (Application) Alternate Selectivity for a Variety of Polar Types, Ideal for Lipids
For Use With (Equipment) Any Flash System