Learn More
Fisherbrand™ Screw Caps for Disposable Glass Tubes
Phenolic caps have cemented-in rubber liners
$286.08 - $688.03
Specifications
Product Type Screw Cap
Liner Material Rubber
Pre-sterilized No
Autoclavable Autoclavable
Sterility Non-sterile
View More Specs
Products 4
Catalog Number Color Closure Size Material Price Quantity & Availability  
Catalog Number Color Closure Size Material Price Quantity & Availability  
1495782A
View Documents Promotion Details
Black 13-415 Phenolic
Case of 1000 Each for $596.46
Only null left
1495936B
View Documents Promotion Details
White 15-415 Polypropylene
Case of 1000 Each for $286.08
Only null left
1495782E
View Documents Promotion Details
Black 18-415 Phenolic
Case of 1000 Each for $688.03
Only null left
1495936A
View Documents Promotion Details
Black 15-415 Phenolic
Case of 1000 Each for $507.31
Only null left
Description
Description
Phenolic caps have cemented-in rubber liners
  • Autoclavable
  • Polypropylene cap is linerless
Specifications
Specifications
Screw Cap
No
Non-sterile
Rubber
Autoclavable