missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Screw Caps for Disposable Glass Tubes

Catalog No. 1495936B
Change view
Click to view available options
Color:
Black
White
Closure Size:
13-415
15-415
18-415
Material:
Phenolic
Polypropylene
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Size Material
1495936B White 15-415 Polypropylene
1495782A Black 13-415 Phenolic
1495782E Black 18-415 Phenolic
1495936A Black 15-415 Phenolic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 1495936B
Edge

Phenolic caps have cemented-in rubber liners

  • Autoclavable
  • Polypropylene cap is linerless

Specifications

Certifications/Compliance ASTM E982
Pre-sterilized No
Product Type Screw Cap
Closure Size 15-415
Liner Material Rubber
Autoclavable Autoclavable
Material Polypropylene
Color White
Sterility Non-sterile
For Use With (Equipment) 14-959-35A, 14-959-35B, 14-959-35AA