missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Hazard Communication Labels
GREENER_CHOICE

Available with five different warning messages and color combinations.

$7.38 - $8.14

Specifications

Format Label
For Use With (Application) Specifies the type of hazard and contains a corresponding visual symbol to augment the warning
Quantity 125/Pk.
Size (English) 0.5 x 4 in.
Size (Metric) 1.3 x 10.2 cm
View More Specs

Products 5
Catalog Number Color Display Text Price Quantity & Availability  
Catalog Number Color Display Text Price Quantity & Availability  
15960
GREENER_CHOICE
View Documents Promotion Details
Black/Orange, Orange CORROSIVE
Each for $7.93
Only null left
 
15961
GREENER_CHOICE
View Documents Promotion Details
Black/Green, Green CONTAMINATED
Each for $7.38
Only null left
 
15962
GREENER_CHOICE
View Documents Promotion Details
Red/Yellow, Yellow FLAMMABLE
Each for $7.38
Only null left
 
15964
GREENER_CHOICE
View Documents Promotion Details
Red/Yellow RADIOACTIVE
Each for $8.14
Only null left
 
15963
GREENER_CHOICE
View Documents Promotion Details
Black/Chrome POISON
Each for $7.38
Only null left
 
Description

Description

  • Continuous adhesive tape
  • Specifies the type of hazard and contains a corresponding visual symbol to augment the warning

Order Info

Label dimensions: 0.50 x 4 in. (1.3 x 10.2cm). Rolls are 500 in. (12.7m) in length.

Specifications

Specifications

Label
125/Pk.
1.3 x 10.2 cm
12.7 m
Specifies the type of hazard and contains a corresponding visual symbol to augment the warning
0.5 x 4 in.
500 in.
Videos
SDS
Documents

Documents