missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Hazard Communication Labels
GREENER_CHOICE

Catalog No. 15961
Change view
Click to view available options
Color:
Black/Chrome
Black/Green, Green
Black/Orange, Orange
Red/Yellow
Red/Yellow, Yellow
Display Text:
CONTAMINATED
CORROSIVE
FLAMMABLE
POISON
RADIOACTIVE
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Display Text
15961 Black/Green, Green CONTAMINATED
15960 Black/Orange, Orange CORROSIVE
15962 Red/Yellow, Yellow FLAMMABLE
15964 Red/Yellow RADIOACTIVE
15963 Black/Chrome POISON
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 15961
Only null left

Available with five different warning messages and color combinations.

  • Continuous adhesive tape
  • Specifies the type of hazard and contains a corresponding visual symbol to augment the warning

Order Info

Label dimensions: 0.50 x 4 in. (1.3 x 10.2cm). Rolls are 500 in. (12.7m) in length.

Specifications

Color Black/Green, Green
Format Label
Product Type Hazard Communication Label
Display Text CONTAMINATED
For Use With (Application) Specifies the type of hazard and contains a corresponding visual symbol to augment the warning
Quantity 125/Pk.
Size (English) 0.5 x 4 in.
Size (Metric) 1.3 x 10.2 cm
Length (English) 500 in.
Length (Metric) 12.7 m