missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Hazard Communication Labels

Available with five different warning messages and color combinations.
Description
- Continuous adhesive tape
- Specifies the type of hazard and contains a corresponding visual symbol to augment the warning
Order Info
Label dimensions: 0.50 x 4 in. (1.3 x 10.2cm). Rolls are 500 in. (12.7m) in length.
Chemical Identifiers
| Label | |
| 125/Pk. | |
| 1.3 x 10.2 cm | |
| 12.7 m |
| Specifies the type of hazard and contains a corresponding visual symbol to augment the warning | |
| 0.5 x 4 in. | |
| 500 in. |
Specifications
| Black/Chrome | |
| Hazard Communication Label | |
| Specifies the type of hazard and contains a corresponding visual symbol to augment the warning | |
| 0.5 x 4 in. | |
| 500 in. |
| Label | |
| POISON | |
| 125/Pk. | |
| 1.3 x 10.2 cm | |
| 12.7 m |