Learn More
Fisherbrand™ Extra-Deep Disposable Petri Dishes
Fisherbrand™ Extra-Deep Disposable Petri Dishes are for fungal cultures and plant propagation.
$741.36 - $854.25
Specifications
Disposable Single-use
Material Polystyrene
Sterility Sterile
For Use With (Application) For fungal cultures and plant propagation
Packaging Sleeve
View More Specs
Products 2
Catalog Number Closure Type Price Quantity & Availability  
Catalog Number Closure Type Price Quantity & Availability  
0875711
View Documents Promotion Details
Vented
Case of 500 Each for $854.25
Only null left
0875711Z
View Documents Promotion Details
Stackable
Case of 500 Each for $741.36
Only null left
Description
Description
  • Disposable polystyrene dishes accommodate deeper media fills for longer culture periods
  • Radiation sterilized
Stackable Dishes
  • 100 × 20mm Dish has a raised straight ridge around the top for stable stacking
Vented Dishes
  • 100 × 25mm Dish features three venting ribs on the underside of the lid
Order Info
Supplied 20 per polyethylene sleeve
Specifications
Specifications
Single-use
Sterile
Sleeve
25 mm
Polystyrene
For fungal cultures and plant propagation
20/Sleeve
100 mm