Learn More
Fisherbrand™ Ultra High Purity Hydrogen Gas Generators Accessory, Preventative Maintenance Kit
For use with Ultra High Purity Hydrogen Gas Generators for GC Carrier and Fuel Gas.
Specifications
For Use With (Application) Gas chromatography, fuel gas, carrier gas, flame ionization detector (GC-FID) flame photometric detector (GC-FPD), nitrogen phosphorous detector carrier gas (NPD), thermal conductivity detector carrier gas (TCD), Electrolytic Conductivity Detector reactio
For Use With (Equipment) Ultra High Purity Hydrogen Gas Generators For GC Carrier and Fuel Gas
View More Specs
Includes:
  • 6 Month Kit: DI Cartridge, and In-line Water filter
  • 24 Month Kit: Water Pump and Service Dongle
  • 60 Month Kit: Replacement Dryer Columns and Service Dongle
Products 3
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
14950650
View Documents Promotion Details
Fisherbrand™
M06.HMD.0001L2352
6 Month PM Kit N/A
N/A
14950651
View Documents Promotion Details
Fisherbrand™
M24.HMD.0001L2352
24 Month PM Kit N/A
N/A
14950652
View Documents Promotion Details
Fisherbrand™
M60.HMD.0001L2352
60 Month PM Kit N/A
N/A
Description
Description
6 month PM Kit
  • Helps ensure quality of deionized water in Fisher Scientifc™ UHP range of H2 generators maximizing life of PEM cell
24 month PM Kit
  • Required at 16,000 hours of operation when Fisher Scientific UHP generator will alarm indicating that service is due
60 month PM Kit
  • Required at 40,000 hours of operation when Fisher Scientific UHP generator will alarm indicating that service is due
Specifications
Specifications
Gas chromatography, fuel gas, carrier gas, flame ionization detector (GC-FID) flame photometric detector (GC-FPD), nitrogen phosphorous detector carrier gas (NPD), thermal conductivity detector carrier gas (TCD), Electrolytic Conductivity Detector reactio
Ultra High Purity Hydrogen Gas Generators For GC Carrier and Fuel Gas