Learn More
Fisherbrand™ Ultra High Purity Hydrogen Gas Generators Accessory, Preventative Maintenance Kit
For use with Ultra High Purity Hydrogen Gas Generators for GC Carrier and Fuel Gas.
Supplier: Fisherbrand™ M24.HMD.0001L2352
Includes: 24 Month Kit: Water Pump and Service Dongle
Description
6 month PM Kit- Helps ensure quality of deionized water in Fisher Scientifc™ UHP range of H2 generators maximizing life of PEM cell
- Required at 16,000 hours of operation when Fisher Scientific UHP generator will alarm indicating that service is due
- Required at 40,000 hours of operation when Fisher Scientific UHP generator will alarm indicating that service is due
Chemical Identifiers
| Gas chromatography, fuel gas, carrier gas, flame ionization detector (GC-FID) flame photometric detector (GC-FPD), nitrogen phosphorous detector carrier gas (NPD), thermal conductivity detector carrier gas (TCD), Electrolytic Conductivity Detector reactio | |
| Ultra High Purity Hydrogen Gas Generators For GC Carrier and Fuel Gas |
Specifications
| 24 Month PM Kit | |
| Gas chromatography, fuel gas, carrier gas, flame ionization detector (GC-FID) flame photometric detector (GC-FPD), nitrogen phosphorous detector carrier gas (NPD), thermal conductivity detector carrier gas (TCD), Electrolytic Conductivity Detector reactio | |
| Replacement float, water pump, service reset tool |
| 24 Momth PM Kit for Fisher Scientific Ultra High Purity H2 Generator | |
| Ultra High Purity Hydrogen Gas Generators For GC Carrier and Fuel Gas |