Learn More
Fisherbrand™ Tissue Path Superfrost™ Plus Gold Slides
Specifically designed for frozen tissue sections
$106.64 - $322.85
Specifications
| Frosted | SuperFrost |
|---|---|
| Type | Superfrost Plus Gold Slide |
| Color | White |
| Coating | Frozen Tissue Adhesion |
Description
Specifically designed for frozen tissue sections
- Made with a revolutionary, new adhesive technology
- Attract and chemically bond fresh or formalin-fixed frozen tissue sections firmly to the slide surface
- Ideal for special stains, immunocytochemical and in situ DNA hybridization techniques when applied to fixed or frozen tissue sections
- Compatible with both toluidine blue and hematoxylin and eosin rapid frozen section stains
Specifications
| SuperFrost | |
| Superfrost Plus Gold Slide | |
| White | |
| Frozen Tissue Adhesion |