Learn More
Fisherbrand™ Tissue Path Superfrost™ Plus Gold Slides
Catalog No. 22035813
Change view
Click to view available options
Quantity:
144/Pk.
25/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22035813 25/Pk.
1518848 144/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22035813
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifically designed for frozen tissue sections
  • Made with a revolutionary, new adhesive technology
  • Attract and chemically bond fresh or formalin-fixed frozen tissue sections firmly to the slide surface
  • Ideal for special stains, immunocytochemical and in situ DNA hybridization techniques when applied to fixed or frozen tissue sections
  • Compatible with both toluidine blue and hematoxylin and eosin rapid frozen section stains
Specifications
Frosted SuperFrost
Type Superfrost Plus Gold Slide
Color White
Coating Frozen Tissue Adhesion
Quantity 25/Pk.