missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-Biosciences First Strand cDNA Synthesis Kit
Description
- First strand cDNA synthesis kit is in a 5x master mix form which contains Reverse Transcriptase (RT), recombinant RNase inhibitor, dNTPs and an optimized buffer with protein stabilizers.
- G-biosciences RT is a recombinant Moloney Murine Leukemia Virus (MMLV) reverse transcriptase with reduced RNase H activity.
- This RT is an RNA-directed DNA polymerase which lacks 3' - 5' exonuclease activity and can produce full-length cDNA from long RNA templates.
Specifications
Specifications
| Detection Method | qPCR, RT-PCR |
| Form | Liquid |
| GC-Rich PCR Performance | High |
| PCR Method | 1-step RT-qPCR, 2-step RT-qPCR, qPCR, Standard PCR, Multiplex qPCR |
| Reaction Speed | Fast or Standard |
| Label or Dye | Biotin, Unconjugated, No ROX, Unlabeled, Biotin and Other Hapten |
| Product Type | Synthesis Kit |
| For Use With (Application) | Gene Expression, Genotyping, Pathogen Detection, Virus Detection, Real Time PCR (qPCR) |
| No. of Reactions | 30 |