Learn More
Thermo Scientific™ Finger Tite Connectors for Thermo Scientific™ and Agilent™ Instruments
Thermo Scientific Finger Tite Connectors for Thermo Scientific and Agilent Instruments offer easier fit, reliability and leak-free connections.
$216.33 - $908.10
Specifications
| For Use With (Equipment) | TRACE 1300 Series GC and Agilent GC Instruments |
|---|
| Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
|
290SA131
|
Thermo Scientific™
290SA131 |
Split/Splitless and MS Starter Kit |
Each for $908.10
|
|
|||||
|
290SA132
|
Thermo Scientific™
290SA132 |
Split/Splitless and FID Starter Kit |
Each for $860.14
|
|
|||||
|
290SA133
|
Thermo Scientific™
290SA133 |
Split/Splitless Injector Base Seal |
Pack of 2 Each for $216.33
|
|
|||||
Description
Thermo Scientific™ Finger Tite Connectors for Thermo Scientific™ and Agilent Instruments offer easier fit, reliability and leak-free connections.
Advantages over conventional graphite/polymer ferrules:
- Perfect metal-to-metal seal eliminates contamination and gives lower air/water background
- No need to retighten ferrules, as they expand and contract with the fitting over GC operating temperatures
- Easy to handle with the nut touchable even with a hot injector/detector
- No tools required
Specifications
| TRACE 1300 Series GC and Agilent GC Instruments |