Learn More
Thermo Scientific™ Finger Tite Connectors for Thermo Scientific™ and Agilent™ Instruments
Catalog No. 290SA131
Change view
Click to view available options
Type:
Split/Splitless and FID Starter Kit
Split/Splitless and MS Starter Kit
Split/Splitless Injector Base Seal
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
290SA131 Split/Splitless and MS Starter Kit
290SA132 Split/Splitless and FID Starter Kit
290SA133 Split/Splitless Injector Base Seal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 290SA131 Supplier Thermo Scientific™ Supplier No. 290SA131
Thermo Scientific Finger Tite Connectors for Thermo Scientific and Agilent Instruments offer easier fit, reliability and leak-free connections.
Thermo Scientific™ Finger Tite Connectors for Thermo Scientific™ and Agilent Instruments offer easier fit, reliability and leak-free connections.
Advantages over conventional graphite/polymer ferrules:
  • Perfect metal-to-metal seal eliminates contamination and gives lower air/water background
  • No need to retighten ferrules, as they expand and contract with the fitting over GC operating temperatures
  • Easy to handle with the nut touchable even with a hot injector/detector
  • No tools required
Specifications
For Use With (Equipment) TRACE 1300 Series GC and Agilent GC Instruments
Description MS Starter Kit; 1/Pk.
Type Split/Splitless and MS Starter Kit
Quantity 1/Pk.