Learn More
Thermo Scientific™ Finger Tite Connectors for Thermo Scientific™ and Agilent™ Instruments
Description
Thermo Scientific™ Finger Tite Connectors for Thermo Scientific™ and Agilent Instruments offer easier fit, reliability and leak-free connections.
Advantages over conventional graphite/polymer ferrules:
- Perfect metal-to-metal seal eliminates contamination and gives lower air/water background
- No need to retighten ferrules, as they expand and contract with the fitting over GC operating temperatures
- Easy to handle with the nut touchable even with a hot injector/detector
- No tools required
Specifications
Specifications
| For Use With (Equipment) | TRACE 1300 Series GC and Agilent GC Instruments |
| Description | Injector/Detector Starter Kit; 1/Pk. |
| Type | Split/Splitless and FID Starter Kit |
| Quantity | 1/Pk. |