missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Finasteride

Catalog No. 329350
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
329350 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 329350 Supplier R&D Systems™ Supplier No. 3293/50

missing translation for 'validMainframeMsg'

A Tocris product, part of R&D Systems. Type II 5α-reductase inhibitor

Antiandrogen that inhibits type II 5α reductase (IC50 = 65 nM). Suppresses the conversion of testosterone to dihydrotestosterone. Reduces prostatic dihydrotestosterone levels and prostate size in vivo. Orally active.

Chemical Identifiers

CAS 98319-26-7
Molecular Formula C23H36N2O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 372.54
Purity >99%
Molecular Weight (g/mol) 372.553
Chemical Name or Material Finasteride
Recommended Storage Store at Room Temperature