Learn More
Thermo Scientific™ Fiberlite™ F14-6 x 250LE Rotor Adapters
Combine this rotor Adapter with the Fiberlite F14-6 x 250LE fixed-angle rotor.
$467.27 - $2882.65
| Catalog Number | Mfr. No. | Capacity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity | Price | Quantity | |||||
|
00-388
|
Thermo Scientific™
00388 |
5 mL Round-bottom Tube |
Each for $467.27
|
|
|||||
|
00-456
|
Thermo Scientific™
00456 |
7 mL Round-bottom Tube |
Each for $990.52
|
|
|||||
|
01-001-38
|
Thermo Scientific™
0100138 |
50 mL Round-bottom Tube (Set of 2) |
Each for $1,163.00
|
|
|||||
|
75-100-136
|
Thermo Scientific™
75100136 |
50 mL Conical Tube (Set of 2) |
Pack of 2 Each for $2,882.65
|
|
|||||
|
75-101-073
|
Thermo Scientific™
75101073 |
15 mL Conical Tube (Set of 2) |
Pack of 2 Each for $2,730.06
|
|
|||||
Description
Combine Thermo Scientific™ Fiberlite™ F14-6 x 250LE Rotor Adapters with the Fiberlite F14-6 x 250LE fixed-angle rotor.
Note:- Adapters 00369 and 00456 must be used together for 7mL round-bottom tubes
Ordering Information:
Fiberlite F14-6x250LE fixed-angle rotor body (75003662) is also available. Contact your sales representative for more information.
Specifications
| Rotor Adapter |