missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fiberlite™ F14-6 x 250LE Rotor Adapters

Catalog No. 75101073
Change view
Click to view available options
Capacity:
15 mL Conical Tube (Set of 2)
5 mL Round-bottom Tube
50 mL Conical Tube (Set of 2)
50 mL Round-bottom Tube (Set of 2)
7 mL Round-bottom Tube
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
75101073 15 mL Conical Tube (Set of 2)
00388 5 mL Round-bottom Tube
00456 7 mL Round-bottom Tube
0100138 50 mL Round-bottom Tube (Set of 2)
75100136 50 mL Conical Tube (Set of 2)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 75101073 Supplier Thermo Scientific™ Supplier No. 75101073

Combine this rotor Adapter with the Fiberlite F14-6 x 250LE fixed-angle rotor.

Combine Thermo Scientific™ Fiberlite™ F14-6 x 250LE Rotor Adapters with the Fiberlite F14-6 x 250LE fixed-angle rotor.

Note:
  • Adapters 00369 and 00456 must be used together for 7mL round-bottom tubes

Ordering Information:

Fiberlite F14-6x250LE fixed-angle rotor body (75003662) is also available. Contact your sales representative for more information.

Specifications

Type Rotor Adapter
Capacity 15 mL Conical Tube (Set of 2)