Learn More
Cytiva Fetal Bovine Serum (U.S.), Super Low IgG
Processed using protein G chromatography to reduce the levels to those required for monoclonal antibody production
$373.69 - $1,678.67
Specifications
Content And Storage 2 to 8°C
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Processing Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Processing Price Quantity & Availability  
SH3089802
View Documents Promotion Details
Cytiva
SH30898.02
100 mL None
Each for $373.69
Only null left
SH3089803
View Documents Promotion Details
Cytiva
SH30898.03
500 mL None
Each for $1,678.67
Only null left
Description
Description
Fetal bovine serum (FBS) has natural levels of IgG that are too high for some cell culture applications. Lots can be prescreened to find the lowest naturally occuring levels, but as a biological product, FBS is frequently in short supply. Even lots with naturally low IgG levels may still contain too much for some applications. Examples of typical IgG levels in bovine serum products can be found in the table below.
  • Performance is equivalent to non-processed FBS
  • Suitable for applications where low levels of specific antibody are required (e.g. propagation of bovine viruses, monoclonal antibody production)
Specifications
Specifications
2 to 8°C