missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Fetal Bovine Serum (U.S.), Super Low IgG

Catalog No. SH3089802
Change view
Click to view available options
Quantity:
100 mL
500 mL
Processing:
None
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3089802 100 mL None
SH3089803 500 mL None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3089802 Supplier Cytiva Supplier No. SH30898.02
Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Processed using protein G chromatography to reduce the levels to those required for monoclonal antibody production

Fetal bovine serum (FBS) has natural levels of IgG that are too high for some cell culture applications. Lots can be prescreened to find the lowest naturally occuring levels, but as a biological product, FBS is frequently in short supply. Even lots with naturally low IgG levels may still contain too much for some applications. Examples of typical IgG levels in bovine serum products can be found in the table below.

  • Performance is equivalent to non-processed FBS
  • Suitable for applications where low levels of specific antibody are required (e.g. propagation of bovine viruses, monoclonal antibody production)

Specifications

Content And Storage 2 to 8°C
Quantity 100 mL
Processing None
Heat Inactivated No