missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Fetal Bovine Serum, Premium, United States Origin
GREENER_CHOICE

Collected at government approved abattoirs in the United States

Supplier:  Corning™ 35015CF

 View more versions of this product

Catalog No. MT35015CF


Only null left

Description

Description

  • True pooled and triple 0.1 micron sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements
  • Granted a Certificate of Suitability, R0-CEP 2018-271, by the European Directorate for the Quality of Medicines (EDQM).
  • Validated process used to heath inactivate serum at 56°C ± 2°C for 30 minutes via bulk inactivation of entire lot in the pooling tank or individual bottles in a water bath.
  • Validated process used to gamma irradiate serum within one of two separate dose ranges: 25-40 kGy and 30-45 kGy.
Specifications

Chemical Identifiers

-40°C to -10°C
United States
United States Origin
Bovine
Liquid

Specifications

-40°C to -10°C
United States
50 mL
United States Origin
Bovine
Liquid
None
No
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 60 months

SDS
Documents

Documents

Promotions

Promotions