missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Fetal Bovine Serum, Premium, United States Origin
GREENER_CHOICE

Catalog No. p-7192941
Change view
Click to view available options
Quantity:
50 mL
500 mL
Serum Treatment:
Heat-inactivated
None
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Serum Treatment
MT35016CF 50 mL Heat-inactivated
MT35015CF 50 mL None
MT35017CV 500 mL Heat-inactivated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MT35016CF Supplier Corning™ Supplier No. 35016CF
Only null left

Collected at government approved abattoirs in the United States

  • True pooled and triple 0.1 micron sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements
  • Granted a Certificate of Suitability, R0-CEP 2018-271, by the European Directorate for the Quality of Medicines (EDQM).
  • Validated process used to heath inactivate serum at 56°C ± 2°C for 30 minutes via bulk inactivation of entire lot in the pooling tank or individual bottles in a water bath.
  • Validated process used to gamma irradiate serum within one of two separate dose ranges: 25-40 kGy and 30-45 kGy.

Specifications

Content And Storage -40°C to -10°C
Species Bovine
Country of Origin United States
Form Liquid
Quantity 50 mL
Serum Treatment Heat-inactivated
Source United States Origin
Heat Inactivated Yes