missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrous Sulfate, Dried Powder, FCC, 86-89%, Spectrum™ Chemical
F1063, 13463-43-9, FeSO4·xH2O
Supplier: Spectrum Chemical Mfg Cor F106325KG
Description
Spectrum™ Chemical Ferrous Sulfate, Dried Powder, FCC is used as an iron fortification food supplement additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 13463-43-9 | |
| 2.5 kg | |
| XBDUTCVQJHJTQZ-UHFFFAOYSA-L | |
| λ²-iron(2+) hydrate sulfate | |
| 86 to 89% | |
| Poly Pail |
| 100% | |
| FeH2O5S | |
| O.[Fe++].[O-]S([O-])(=O)=O | |
| 169.92 | |
| FCC |
Chemical Identifiers
| 13463-43-9 | |
| 169.92 | |
| λ²-iron(2+) hydrate sulfate |
| FeH2O5S | |
| XBDUTCVQJHJTQZ-UHFFFAOYSA-L | |
| O.[Fe++].[O-]S([O-])(=O)=O |