missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Sulfate, Dried Powder, FCC, 86-89%, Spectrum™ Chemical

Catalog No. p-7157428
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602707 500 g Poly Bottle
18602708 2.5 kg Poly Pail
18602709 12 kg Poly Pail
18602710 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602707 Supplier Spectrum Chemical Mfg Cor Supplier No. F1063500GM
Only null left

F1063, 13463-43-9, FeSO4·xH2O

Spectrum™ Chemical Ferrous Sulfate, Dried Powder, FCC is used as an iron fortification food supplement additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 13463-43-9
Molecular Formula FeH2O5S
Molecular Weight (g/mol) 169.92
InChI Key XBDUTCVQJHJTQZ-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) hydrate sulfate
SMILES O.[Fe++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 86 to 89%
Grade FCC
Packaging Poly Bottle