missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrous Gluconate, FCC, 97-102%, Spectrum™ Chemical
F1053, 12389-15-0, C12H22FeO14·2H2O
Supplier: Spectrum Chemical Mfg Cor F105312KG
Description
Spectrum™ Chemical Ferrous Gluconate, FCC is used as an iron fortification food supplement additive. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 12389-15-0 | |
| 12 kg | |
| OKGNXSFAYMSVNN-UHFFFAOYNA-L | |
| λ²-iron(2+) bis(2,3,4,5,6-pentahydroxyhexanoate) dihydrate | |
| 97 to 102% | |
| 6.5-10% |
| 100% | |
| C12H26FeO16 | |
| O.O.[Fe++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O | |
| 482.17 | |
| FCC | |
| Poly Pail |
Chemical Identifiers
| 12389-15-0 | |
| 482.17 | |
| λ²-iron(2+) bis(2,3,4,5,6-pentahydroxyhexanoate) dihydrate |
| C12H26FeO16 | |
| OKGNXSFAYMSVNN-UHFFFAOYNA-L | |
| O.O.[Fe++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O |