missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Gluconate, FCC, 97-102%, Spectrum™ Chemical

Catalog No. p-7157423
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602699 500 g Amber Glass Bottle
18602700 2.5 kg Poly Pail
18602701 12 kg Poly Pail
18602702 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602699 Supplier Spectrum Chemical Mfg Cor Supplier No. F1053500GM
Only null left

F1053, 12389-15-0, C12H22FeO14·2H2O

Spectrum™ Chemical Ferrous Gluconate, FCC is used as an iron fortification food supplement additive. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 12389-15-0
Molecular Formula C12H26FeO16
Molecular Weight (g/mol) 482.17
InChI Key OKGNXSFAYMSVNN-UHFFFAOYNA-L
IUPAC Name λ²-iron(2+) bis(2,3,4,5,6-pentahydroxyhexanoate) dihydrate
SMILES O.O.[Fe++].OCC(O)C(O)C(O)C(O)C([O-])=O.OCC(O)C(O)C(O)C(O)C([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 97 to 102%
Grade FCC
Loss on Drying 6.5-10%
Packaging Amber Glass Bottle