missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrous Ammonium Sulfate, Hexahydrate, Crystal, ACS, 98.5-101.5%, Spectrum™ Chemical
$224.88 - $15435.12
Chemical Identifiers
| CAS | 7783-85-9 |
|---|---|
| Molecular Formula | FeH20N2O14S2 |
| Molecular Weight (g/mol) | 392.13 |
| MDL Number | MFCD00150530 |
| InChI Key | MQLVWQSVRZVNIP-UHFFFAOYSA-L |
| IUPAC Name | λ²-iron(2+) diammonium hexahydrate disulfate |
| SMILES | [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18610557
|
Spectrum Chemical Mfg Cor
F105025GM |
25 g | Amber Glass Bottle |
Each for $224.88
|
|
||||
|
18610558
|
Spectrum Chemical Mfg Cor
F1050125GM |
125 g | Amber Glass Bottle |
Each for $443.38
|
|
||||
|
18610559
|
Spectrum Chemical Mfg Cor
F1050500GM |
500 g | Amber Glass Bottle |
Each for $934.21
|
|
||||
|
18610560
|
Spectrum Chemical Mfg Cor
F105025KG |
2.5 kg | Amber Glass Bottle |
Each for $3,751.15
|
|
||||
|
18610561
|
Spectrum Chemical Mfg Cor
F105012KG |
12 kg | Poly Pail |
Each for $4,067.12
|
|
||||
|
18610562
|
Spectrum Chemical Mfg Cor
F105050KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 7783-85-9 | |
| 392.13 | |
| MQLVWQSVRZVNIP-UHFFFAOYSA-L | |
| [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
| FeH20N2O14S2 | |
| MFCD00150530 | |
| λ²-iron(2+) diammonium hexahydrate disulfate |
Specifications
| 7783-85-9 | |
| 25 g | |
| MFCD00150530 | |
| [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
| 392.13 | |
| ACS |
| 1 | |
| FeH20N2O14S2 | |
| MQLVWQSVRZVNIP-UHFFFAOYSA-L | |
| λ²-iron(2+) diammonium hexahydrate disulfate | |
| 98.5 to 101.5% | |
| Amber Glass Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.