missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferrous Ammonium Sulfate, Hexahydrate, Crystal, ACS, 98.5-101.5%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor F1050125GM
Specifications
| 7783-85-9 | |
| 125 g | |
| MFCD00150530 | |
| [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
| 392.13 | |
| ACS |
| 1 | |
| FeH20N2O14S2 | |
| MQLVWQSVRZVNIP-UHFFFAOYSA-L | |
| λ²-iron(2+) diammonium hexahydrate disulfate | |
| 98.5 to 101.5% | |
| Amber Glass Bottle |
Chemical Identifiers
| 7783-85-9 | |
| 392.13 | |
| MQLVWQSVRZVNIP-UHFFFAOYSA-L | |
| [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
| FeH20N2O14S2 | |
| MFCD00150530 | |
| λ²-iron(2+) diammonium hexahydrate disulfate |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.