missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Ammonium Sulfate, Hexahydrate, Crystal, ACS, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18610557
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610557 25 g Amber Glass Bottle
18610558 125 g Amber Glass Bottle
18610559 500 g Amber Glass Bottle
18610560 2.5 kg Amber Glass Bottle
18610561 12 kg Poly Pail
18610562 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18610557 Supplier Spectrum Chemical Mfg Cor Supplier No. F105025GM
Only null left

Chemical Identifiers

CAS 7783-85-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 98.5 to 101.5%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.