missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Ammonium Sulfate, 0.250 N (N/4), 0.250 M (M/4), Ricca Chemical

Catalog No. p-3251686
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R3150000100 100 mL Amber Glass Bottle
315032 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R3150000100 Supplier Ricca Chemical Company Supplier No. R3150000100C
Only null left

Chemical Identifiers

CAS 7783-85-9 , 7732-18-5 , 7664-93-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
Synonym Ammonium Iron (II) Sulfate, Iron (II) Ammonium Sulfate Hexahydrate, Iron(II) ammonium sulfate
PubChem CID 115148
IUPAC Name λ2-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Green
CAS Min % 8.48, 86.0, 3.18
CAS Max % 8.82, 90.0, 3.31
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 100 mL
Physical Form Liquid
Concentration or Composition Notes 0.250Normal,0.250Molar
Chemical Name or Material Ferrous Ammonium Sulfate