missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Ammonium Sulfate, 0.100 N (N/10), 0.100 M (M/10), Ricca Chemical

Catalog No. p-3251685
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
10 L
Packaging:
Amber Glass Bottle
Cubitainer
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R3145000100 100 mL Amber Glass Bottle
314516 500 mL Amber Glass Bottle
314532 1 L Amber Glass Bottle
31451 4 L Amber Glass Bottle
314525 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. R3145000100 Supplier Ricca Chemical Company Supplier No. R3145000100C
Only null left

Chemical Identifiers

CAS 7783-85-9 , 7732-18-5 , 7664-93-9
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
Synonym Ammonium Iron (II) Sulfate, Iron (II) Ammonium Sulfate Hexahydrate, Iron(II) ammonium sulfate
PubChem CID 115148
IUPAC Name λ2-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color Green
CAS Min % 3.57, 91.0, 3.35
CAS Max % 3.72, 95.0, 3.49
Grade Laboratory
Packaging Amber Glass Bottle
Quantity 100 mL
Physical Form Liquid
Concentration or Composition Notes 0.100Normal,0.100Molar
Chemical Name or Material Ferrous Ammonium Sulfate