missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Ferrocene | |
| 176°C | |
| C10H10Fe | |
| 25 g | |
| ferrocene, bis cyclopentadienyl iron | |
| [Fe].c1cccc1.c1cccc1 | |
| 186.04 | |
| 186.04 | |
| Crystalline Powder |
| 102-54-5 | |
| 249°C | |
| MFCD00001427 | |
| 1325 | |
| DFRHTHSZMBROSH-UHFFFAOYSA-N | |
| Ferrocene | |
| 25199998 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 102-54-5 | |
| 186.04 | |
| DFRHTHSZMBROSH-UHFFFAOYSA-N | |
| 25199998 | |
| [Fe].c1cccc1.c1cccc1 |
| C10H10Fe | |
| MFCD00001427 | |
| ferrocene, bis cyclopentadienyl iron | |
| Ferrocene |
Safety and Handling
EINECSNumber : (3)-2568
RTECSNumber : LK0700000
TSCA : Yes