accessibility menu, dialog, popup

UserName

Fe

CAS RN 102-54-5

IUPAC Name: Ferrocene
Synonyms: —
Molecular Weight (g/mol): 186.04
Molecular Formula: C10H10Fe
InChi Key: DFRHTHSZMBROSH-UHFFFAOYSA-N
SMILES: [Fe].c1cccc1.c1cccc1
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)