missing translation for 'onlineSavingsMsg'
Learn More

Ferrocene 98.0+%, TCI America™

Catalog No. p-7126217
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D044425G 25 g
D0444100G 100 g
D0444500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D044425G Supplier TCI America Supplier No. D044425G

Chemical Identifiers

CAS 102-54-5
Molecular Formula C10H10Fe
Molecular Weight (g/mol) 186.04
MDL Number MFCD00001427
InChI Key DFRHTHSZMBROSH-UHFFFAOYSA-N
Synonym ferrocene, bis cyclopentadienyl iron
PubChem CID 25199998
IUPAC Name Ferrocene
SMILES [Fe].c1cccc1.c1cccc1

Specifications

Melting Point 176°C
Boiling Point 249°C
Quantity 25 g
UN Number 1325
Formula Weight 186.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ferrocene
EINECSNumber (3)-2568
RTECSNumber LK0700000
TSCA Yes