missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ferric Chloride, MP Biomedicals
Manufacturer: Mp Biomedicals 0215349980
Specifications
Ferric chloride, anhydrous | |
7705-08-0 | |
100g | |
MFCD00011005 | |
162.21 | |
2.898g/cm3 at 25°C |
Chemistry | |
Black | |
FeCl3 | |
Chlorure Perrique, Iron (III) Chloride | |
Powder |
Chemical Identifiers
7705-08-0 | |
FeCl3 | |
MFCD00011005 | |
Chlorure Perrique, Iron (III) Chloride |
Safety and Handling
EINECSNumber : 231-096-4
storageNote1 : Store desiccated. Store under nitrogen
Recommended Storage : Store at Room Temperature (15° to 30°C)