missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride, MP Biomedicals

Catalog No. p-3547101
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15349980 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15349980 Supplier MP Biomedicals Inc Supplier No. 0215349980
Only null left

Chemical Identifiers

CAS 7705-08-0
Molecular Formula Cl3Fe
Molecular Weight (g/mol) 162.20
MDL Number MFCD00011005
InChI Key RBTARNINKXHZNM-UHFFFAOYSA-K
Synonym Chlorure Perrique, Iron (III) Chloride
IUPAC Name iron(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Color Black
Physical Form Powder
Quantity 100 g
Formula Weight 162.21
Density 2.898g/cm3 at 25°C
Chemical Name or Material Ferric chloride, anhydrous, Chemistry
EINECSNumber 231-096-4
storageNote1 Store desiccated. Store under nitrogen
Recommended Storage Store at Room Temperature (15° to 30°C)