missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Chloride, 10% (w/v), Ricca Chemical
$68.94 - $589.69
Chemical Identifiers
| CAS | 10025-77-1 |
|---|---|
| Molecular Formula | Cl3FeH12O6 |
| Molecular Weight (g/mol) | 270.29 |
| MDL Number | MFCD00149712 |
| InChI Key | NQXWGWZJXJUMQB-UHFFFAOYSA-K |
| Synonym | Ferric chloride hexahydrate, Iron Trichloride, Perchlorure de Fer |
| PubChem CID | 24380 |
| ChEBI | CHEBI:30808 |
| IUPAC Name | iron(3+) hexahydrate trichloride |
| SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
31204
|
Ricca Chemical Company
31204 |
120 mL | Natural Poly Bottle |
Each for $68.94
|
|
||||
|
3120-16
|
Ricca Chemical Company
312016 |
500 mL | Natural Poly Bottle |
Each for $112.59
|
|
||||
|
3120-32
|
Ricca Chemical Company
312032 |
1 L | Natural Poly Bottle |
Each for $181.72
|
|
||||
|
3120-1
|
Ricca Chemical Company
31201 |
4 L | Natural Poly Bottle |
Each for $589.69
|
|
||||
Chemical Identifiers
| 10025-77-1 | |
| 270.29 | |
| NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
| 24380 | |
| iron(3+) hexahydrate trichloride |
| Cl3FeH12O6 | |
| MFCD00149712 | |
| Ferric chloride hexahydrate, Iron Trichloride, Perchlorure de Fer | |
| CHEBI:30808 | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
Specifications
| 10025-77-1,7732-18-5,7647-01-0 | |
| 9.18,92.0,1.08 | |
| MFCD00149712 | |
| NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
| iron(3+) hexahydrate trichloride | |
| 24380 | |
| 10% | |
| 10% (w/v) Aqueous Solution | |
| Acidic | |
| Ferric Chloride Solution |
| 8.82,88.0,1.04 | |
| Cl3FeH12O6 | |
| Ferric chloride hexahydrate, Iron Trichloride, Perchlorure de Fer | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] | |
| 270.29 | |
| CHEBI:30808 | |
| Laboratory | |
| Natural Poly Bottle | |
| 120 mL |