missing translation for 'onlineSavingsMsg'
Learn More

Ferric Ammonium Sulfate, For Chloride (Volhard's Indicator), Certified, 8%, LabChem™

Catalog No. LC142702
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC142702 1 L Poly Bottle
LC142701 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC142702 Supplier LabChem Supplier No. LC142702

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7783-83-7 , 7732-18-5
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.176
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

Specifications

CAS Max % 92, 8
Identification Passes Test
Packaging Poly Bottle
Suitability Passes Test
Linear Formula FeNH4S2O8·12H2O
Decomposition Information Sulfur compounds; Gaseous Ammonia; Nitrogen oxides
Color Brown/Yellow
Solubility Information Soluble in water
Quantity 1 L
Physical Form Liquid
Formula Weight 482.19
Percent Purity 8%
Grade Certified
Chemical Name or Material Ferric Ammonium Sulfate, For Chloride (Volhard's Indicator)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature