missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Ammonium Sulfate, 3% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company R305600020F
Specifications
| Ferric Ammonium Sulfate | |
| 7783-83-7 , 7732-18-5 | |
| 3% (w/v) | |
| ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g | |
| [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] | |
| 482.176 | |
| Odorless |
| Aqueous Solution | |
| 20 L | |
| FeH28NO20S2 | |
| LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
| azanium;iron(3+);disulfate;dodecahydrate | |
| 197097 | |
| Cubitainer |
Chemical Identifiers
| 7783-83-7 | |
| 482.176 | |
| ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g | |
| azanium;iron(3+);disulfate;dodecahydrate |
| FeH28NO20S2 | |
| LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
| 197097 | |
| [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] |
Safety and Handling
Recommended Storage : 15°C - 30°C (59°F - 86°F)