missing translation for 'onlineSavingsMsg'
Learn More

Ferric Ammonium Sulfate, 0.0500 N, Ricca Chemical

Supplier:  Ricca Chemical Company R30590004A

Quantity 4 L
Packaging Natural Poly Bottle

 View more versions of this product

Catalog No. R30590004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

7783-83-7 , 7732-18-5 , 7664-93-9
482.18
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
iron(3+) ammonium dodecahydrate disulfate
FeH28NO20S2
LCPUDZUWZDSKMX-UHFFFAOYSA-K
197097  
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Ferric Ammonium Sulfate
7783-83-7 , 7732-18-5 , 7664-93-9
FeH28NO20S2
LCPUDZUWZDSKMX-UHFFFAOYSA-K
iron(3+) ammonium dodecahydrate disulfate
197097  
Aqueous Solution
0.0500 Normal
0.05000 ± 0.00005 N at 20°C
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
482.18
Natural Poly Bottle
4 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents