Learn More
Thermo Scientific™ X-Gluc
Catalog No. ferr0851
Change view
Click to view available options
Quantity:
0.1 g
Catalog No. Quantity
FERR0851 0.1 g
1 options
Catalog No. FERR0851 Supplier Thermo Scientific™ Supplier No. R0851
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Effectively screen colonies with X-Gluc, a substrate for ß-glucuronidase (GUS) which is encoded by gusA, a widely used reporter gene.
X-Gluc (5-bromo-4-chloro-3-indolyl-beta-D-glucuronic acid, cyclohexylammonium salt) is a substrate for beta-glucuronidase (GUS) which is encoded by gusA, a widely used reporter gene. Glucuronidase cleaves X-Gluc to produce colorless glucuronic acid and an intense blue precipitate of chloro-bromoindigo (see the scheme below).
Glucuronidase cleaves X-Gluc to produce colorless glucuronic acid and an intense blue precipitate of chloro-bromoindigo (see the scheme below).
Recommended for:
  • Detection of GUS expression in plant cells and tissues (1)
  • Detection of infections caused by E.coli
  • Detection of bacterial contamination in food and water samples
Note:
Preparation of a 0.1M stock solution in dimethylformamide or dimethylsulfoxide is recommended.
Specifications
Detection Method Colorimetric
Product Type X-Gluc
Quantity 0.1 g
Substrate Properties Chemical Substrate
Substrate Type Beta-Glucuronidase (GUS) Substrate
For Research Use Only. Not for use in diagnostic procedures.