missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Uracil-DNA Glycosylase (1 U/μL)

Catalog No. FEREN0361
Change view
Click to view available options
Quantity:
200 units
5 x 200 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREN0361 200 units
FEREN0362 5 x 200 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEREN0361 Supplier Thermo Scientific™ Supplier No. EN0361

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Uracil-DNA Glycosylase (UDG, UNG) catalyzes the hydrolysis of the N-glycosylic bond between uracil and sugar in DNA and prevents carry-over of DNA in PCR reactions.

Uracil-DNA Glycosylase (UDG, UNG) catalyzes the hydrolysis of the N-glycosylic bond between uracil and sugar, leaving an apyrimidinic site in uracil-containing single or double-stranded DNA. The enzyme shows no activity on RNA.

  • Active in Fermentas buffers for restriction enzymes and thermophilic polymerases
  • Source: E. coli K12 cells
  • Molecular Weight: 25.6 kDa monomer
Quality Control:
  • The absence of endo-, exodeoxyribonucleases, phosphatases and ribonucleases confirmed by appropriate quality tests

Source:

  • E.coli K12 cells

Molecular Weight:

  • 25.6kDa monomer

Definition of Activity Unit:

  • One unit of the enzyme catalyzes the release 1 nanomole of uracil from uracil-containing DNA template in 60 min. at 37°C
Storage Buffer: The enzyme is supplied in:
  • 30mM Tris-HCl (pH 7.5), 150mM NaCl, 1mM EDTA, 1mM DTT, 0.05% (v/v) Tween 20 and 50% (v/v) glycerol

10X Reaction Buffer:

  • 200mM Tris-HCl (pH 8.2 at 25°C), 10mM EDTA, 100mM NaCl
Inhibition and Inactivation:
  • Inhibitors: Ugi protein from the Bacillus subtilis phage PBS2, protein p56 from the Bacillus subtilis phage phi29 (7).
  • Inactivated by heating at 95°C for 10 min. Enzyme activity is partially restored at temperatures lower than 55°C. Therefore put PCR products on ice after PCR and load directly on a gel

Recommended for:

Control of carry-over contamination in PCR (2); Glycosylase mediated single nucleotide polymorphism detection (GMPD) (3); Site-directed mutagenesis (4); As a probe for protein-DNA interaction studies (5); SNP genotyping; Cloning of PCR products (6); Generation of single strand overhangs of PCR products and cDNA

Note:

The abasic sites formed in DNA by Uracil-DNA Glycosylase may be subsequently cleaved by heat, alkali-treatment or endonucleases that cleave specifically at abasic sites.UDG (UNG) is active in the presence or absence of divalent cations.

Specifications

Concentration 1 U/μL
Content And Storage Uracil-DNA glycolase (1 U/μL) and 10X Reaction Buffer. Store at -20°C.
Enzyme Uracil-DNA Glycosylase
Compatible Buffer 10X Reaction Buffer
Quantity 200 units
Product Type Uracil DNA Glycosylase
What is UDG?

UDG (uracil-DNA-glycosylase) refers to a superfamily of enzymes comprising six sub-families. Family I UDG enzymes are called UNG, after the uracil-N-glycosylase gene. The terms UDG and UNG are commonly used interchangeably because they perform the same function in qPCR, namely, to remove Uracil from dU-containing DNA to prevent carryover contamination from previous PCRs. See the following link: https://www.thermofisher.com/uk/en/home/life-science/pcr/real-time-pcr/real-time-pcr-learning-center/real-time-pcr-basics/what-is-ung-udg.html

For Research Use Only. Not for use in diagnostic procedures.