Learn More
Thermo Scientific™ T7 RNA Polymerase (20 U/μL)
Thermo Scientific™ T7 RNA Polymerase is a DNA-dependent RNA polymerase with strict specificity for its double-stranded promoter.
$161.75 - $491.90
Specifications
Polymerase T7 RNA Polymerase
Concentration 20U/µL
View More Specs
Products 2
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Concentration Quantity Price Quantity & Availability  
FEREP0111
View Documents Promotion Details
Thermo Scientific™
EP0111
20U/µL 5,000 units
Each for $161.75
Only null left
FEREP0112
View Documents Promotion Details
Thermo Scientific™
EP0112
20U/µL 5 x 5,000 units
Each for $491.90
Only null left
Description
Description
Thermo Scientific™ Bacteriophage T7 RNA polymerase is DNA-dependent RNA polymerase with strict specificity its respective double-stranded promoter.
It catalyzes the 5'=>3' synthesis of RNA on either single-stranded DNA or double-stranded DNA downstream from it promoter.
  • Incorporates modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)
Recommended for:
Synthesis of unlabeled and labeled RNA that can be used:for hybridization, in vitro RNA translation; as aRNA, siRNA, substrate in RNase protection assays, template for genomic DNA sequencing.
Note:
Consensus promoter sequences: TAATACGACTCACTATA G GGAGA. The position in bold (+1) indicates the first nucleotide incorporated into RNA during transcription. Only bases at this position through +3 are critical for transcription, and they must be a G and a purine base, respectively.
Specifications
Specifications
T7 RNA Polymerase
20U/µL
Videos
SDS
Documents
Documents
For Research Use Only. Not for use in diagnostic procedures.